Phosphorous acid, 98+%, Thermo Scientific Chemicals
Phosphorous acid, 98+%, Thermo Scientific Chemicals
Phosphorous acid, 98+%, Thermo Scientific Chemicals
Phosphorous acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorous acid, 98+%, Thermo Scientific Chemicals
CAS: 13598-36-2 | H3PO3
Have Questions?
Catalog NumberQuantity
ALFA11189.30250 g
Catalog number ALFA11189.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS95-96-5
IUPAC Name3,6-dimethyl-1,4-dioxane-2,5-dione
Molecular FormulaC6H8O4
InChI KeyJJTUDXZGHPGLLC-UHFFFAOYNA-N
SMILESCC1OC(=O)C(C)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=98.5 %
Melting point122°C to 126°C
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.
Solubility
Soluble in water and alcohol.
Notes
Air sensitive. Hygroscopic. Incompatible with strong bases, oxidizing agents and metals.
RUO – Research Use Only
General References:
  1. Prokop, M.; Bystron, T.; Bouzek, K. Electrochemistry of phosphorus and Hypophosphorus Acid on a Pt electrode. Electrochim. Acta 2015, 160, 214-218.
  2. Grun, A.; Kovacs, R.; Illes Nagy, D.; Garadnay, S.; Greiner, I.; Keglevich, G. Efficient Synthesis of Benzidronate Applying of Phosphorus Trichloride and phosphorus Acid. Lett Drug Des Discov 2015, 12 (1), 78-84.