Antimony(III) oxide, 99%, Thermo Scientific Chemicals
Antimony(III) oxide, 99%, Thermo Scientific Chemicals
Antimony(III) oxide, 99%, Thermo Scientific Chemicals
Antimony(III) oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antimony(III) oxide, 99%, Thermo Scientific Chemicals
CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA11123.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point1550°C (sublimation)
Chemical Name or MaterialAntimony (III) oxide
Melting Point656°C
CAS1309-64-4
Health Hazard 1H351
View more
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Jay A. Young.Antimony(III) Oxide. J. Chem. Educ. 2007, 84 (10),1602.
  2. Nelson Belzile,; Yu-Wei Chen1,; Zijian Wang.Oxidation of antimony (III) by amorphous iron and manganese oxyhydroxides. Chemical Geology. 2001, 174 (4),379-387.