p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA10946.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS305-84-0
IUPAC Name(2S)-2-(3-aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid
Molecular FormulaC9H14N4O3
InChI KeyCQOVPNPJLQNMDC-ZETCQYMHSA-N
SMILESNCCC(=O)N[C@@H](CC1=CN=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation+20.0° to +22.0° (20°C, 589 nm) (c=2, water)
Water=<1 %
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.
Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only
General References:
  1. Radja, I.; Djelad, H.; Morallon, E.; Benyoucef, A. Characterization and electrochemical properties of conducting nanocomposites synthesized from p-anisidine and aniline with titanium carbide by chemical oxidative method. Synth. Met. 2015, 202, 25-32.
  2. Sánchez-Machado, D. I.; López-Cervantes, J.; Núñez-Gastélum, J. A.; de la Mora-López, G. S.; López-Hernández, J.; Paseiro-Losada, P. Effect of the refining process on Moringa oleifera seed oil quality. Food Chem. 2015, 187, 53-57.