Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals
Sodium 1-heptanesulfonate, 99% (dry wt.), water &lt;2%, Thermo Scientific Chemicals
Sodium 1-heptanesulfonate, 99% (dry wt.), water &lt;2%, Thermo Scientific Chemicals
Sodium 1-heptanesulfonate, 99% (dry wt.), water &lt;2%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals
CAS: 22767-50-6 | C7H15NaO3S | 202.244 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA10917.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6381-59-5
IUPAC Namepotassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular FormulaC4H12KNaO10
InChI KeyVZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILESO.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Sulfate (SO4)=<0.005 %
Ammonium (NH4)=<0.002 %
Calcium (Ca)=<0.005 %
View more
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kim, H. Y.; Shin, I. S. Comparison of paralytic shellfish toxin profiles of Alexandrium tamarense and blue mussel (Mytilus edulis) in Korea. Food Sci. Biotechnol. 2015, 24 (2), 751-756.
  2. Chemmalil, L.; Suravajjala, S.; See, K.; Jordan, E.; Furtado, M.; Sun, C.; Hosselet, S. A Novel Approach for Quantitation of Nonderivatized Sialic Acid in Protein Therapeutics Using Hydrophilic Interaction Chromatographic Separation and Nano Quantity Analyte Detection. J. Pharm. Sci. 2015, 104 (1), 15-24.