Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
CAS: 7784-24-9 | AlH24KO20S2 | 474.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
Catalog number ALFA10906.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS244193-52-0
IUPAC Name3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular FormulaC12H23BF4N2
InChI KeyGXZCAMSPWNHTAE-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormViscous liquid
Identification (FTIR)Conforms
Residual halide≤100 ppm (by Ion Chromatography)
Ion Chromatography1-Methyl-3-octylimidazolium : ≥98.5%