4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
CAS: 589-15-1 | C7H6Br2 | 249.933 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA10892.14
Catalog number ALFA10892.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight249.94
Unit Size
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.
Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
RUO – Research Use Only
Gilles Brackmana.; Ulrik Hillaertb.; Serge Van Calenberghb.; Hans J. Nelisa.;Tom Coenyea. Use of quorum sensing inhibitors to interfere with biofilm formation and development in Burkholderia multivorans and Burkholderia cenocepacia. Research in Microbiology. 2009, 160, (2), 144-151.