2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, C19H15ClN4, CAS Number-298-96-4, triphenyltetrazolium chloride, tetrazolium chloride, urocheck, tetrazolium red, tetrazolium chloride, uroscreen, red tetrazolium, vitastain, 2,3,5-triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA10870.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2, 3, 5-Triphenyl-2H-tetrazolium chloride
CAS298-96-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for detection of reducing sugars.
  2. Oxidizing agent, reduced to the colored formazan, capable of introducing a -hydroxyl substituent into steroidal enones: J. Chem. Soc., Perkin 1, 2687 (1988).
  3. McLaughlin, M. R.; Balaa, M. F. Enhanced contrast of bacteriophage plaques in Salmonella with ferric ammonium citrate and sodium thiosulfate (FACST) and tetrazolium red (TZR). J. Microbiol. Methods 2006, 65 (2), 318-323.
  4. Li, X.; Deng, S.; Fu, H. Synergism between red tetrazolium and uracil on the corrosion of cold rolled steel in H2SO4 solution. Corros. Sci. 2009, 51 (6), 1344-1355.