N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals
CAS: 111-41-1 | C4H12N2O | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFA10833.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS90-94-8
IUPAC Name4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
Molecular FormulaC17H20N2O
InChI KeyVVBLNCFGVYUYGU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with HClO4>=97.5 %
Melting point171°C to 176°C (Dec)
Infrared spectrumConforms
Appearance (Color)Blue to dark blue to grey
View more
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.
Solubility
Fully miscible in water.
Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.
RUO – Research Use Only
General References:
  1. Hassan Ali Zamani.; Mohammad Taghi Hamed-Mosavian.;Elham Hamidfar.; Mohammad Reza Ganjali.; Parviz Norouzi. A novel iron (III)-PVC membrane potentiomeric sensor based on N-(2-hydroxyethyl)ethylenediamine-N,N',N″-triacetic acid. Materials Science and Engineering: C. 2008, 28, (8), 1551-1555.
  2. Hiroshi Ogino.; Mamoru Shimoi.; Yoko Saito. Structural identification of the reactive vanadium(III) intermediate formed in the electron-transfer reactions of [N'-(2-hydroxyethyl)ethylenediamine-N,N,N'-triacetato]aquavanadium(III) complex ([V(hedtra)(H2O)]) with halopentaamminecobalt(III) complexes: x-ray crystal structures of [V(hedtra)(H2O)].cntdot.2H2O and K[VO(hedtra)].cntdot.H2O. norg. Chem 1989, 28, (18), 3596-3600.