Ruthenium(IV) oxide, anhydrous, 99.9%
Ruthenium(IV) oxide, anhydrous, 99.9%
Ruthenium(IV) oxide, anhydrous, 99.9%
Ruthenium(IV) oxide, anhydrous, 99.9%
Thermo Scientific Chemicals
Ruthenium(IV) oxide, anhydrous, 99.9%
CAS: 12036-10-1 | O2Ru | 133.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10816.1425 g
Catalog number ALFA10816.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1122-62-9
IUPAC Name1-(pyridin-2-yl)ethan-1-one
Molecular FormulaC7H7NO
InChI KeyAJKVQEKCUACUMD-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5200 to 1.5250 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to yellow to light brown
Infrared spectrumConforms
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Applied as an electrochemical super capacitor material, has high capacity to store charge.
  • Used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, used as active material in super capacitors.
  • Dispersion property of RuO2; used to increase the capacitance of nanocomposite electrode materials for supercapacitors
  • Extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries
RUO – Research Use Only