3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA10723.065 g
Catalog number ALFA10723.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS19614-12-1
IUPAC Name1-bromo-2-(bromomethyl)-4-methoxybenzene
Molecular FormulaC8H8Br2O
InChI KeyMURVUTUZSUEIGI-UHFFFAOYSA-N
SMILESCOC1=CC(CBr)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%