Ethylenediaminetetraacetic acid, 99%
Ethylenediaminetetraacetic acid, 99%
Ethylenediaminetetraacetic acid, 99%
Ethylenediaminetetraacetic acid, 99%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, 99%
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10713.30250 g
Catalog number ALFA10713.30
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS24677-78-9
IUPAC Name2,3-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow-green or cream to brown or grey
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Clarity5% w/v in methanol: clear solution
Melting Point (clear melt)103-109?C
View more
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.
Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.
Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.
RUO – Research Use Only