Lead(II) chromate, 98%, Thermo Scientific Chemicals
Lead(II) chromate, 98%, Thermo Scientific Chemicals
Lead(II) chromate, 98%, Thermo Scientific Chemicals
Lead(II) chromate, 98%, Thermo Scientific Chemicals
Lead(II) chromate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALFA10678.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLead (II) chromate
Melting Point844°C
CAS7758-97-6
Health Hazard 1H350-H360Df-H373
Health Hazard 2GHS H Statement
H350-H360-H373-H201
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.