2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals
2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,5-Diphenyloxazole, 99%, Thermo Scientific Chemicals
CAS: 92-71-7 | C15H11NO | 221.26 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10654.1425 g
Catalog number ALFA10654.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1577-22-6
IUPAC Namehex-5-enoic acid
Molecular FormulaC6H10O2
InChI KeyXUDOZULIAWNMIU-UHFFFAOYSA-N
SMILESOC(=O)CCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
GC>=97.5 %