Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals
CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10510.22100 g
Catalog number ALFA10510.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS9000-01-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<15.0 % (1 g, 105°C)
Heavy metals (as Pb)=<40 ppm
Arsenic (As)=<3 ppm
Infrared spectrumConforms
View more
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.
Solubility
Soluble in hydrochloric acid, nitric acid, diethyl ether, ethyl acetate and acetone. Insoluble in alcohol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Lewis acid catalyst which promotes the rearrangement of acetanilides to o-amino ketones: Tetrahedron Lett., 3217 (1976).
  2. Silyl enol ethers can be ɑ-acylated with acyl chlorides in the presence of a catalytic amount of BiCl3 and NaI or ZnI2; other Lewis acids such as ZnCl2 were less effective: J. Org. Chem., 61, 3885 (1996).
  3. Catalyzes the addition of 1,3-dicarbonyl compounds to Michael acceptors under solvent-free conditions with microwave irradiation: Tetrahedron Lett., 38, 1449 (1997).
  4. For catalysis of Diels-Alder and hetero Diels-Alder reactions, see: J. Org. Chem., 62, 4880 (1997); Tetrahedron Lett., 39, 1161 (1998).
  5. Arenes undergo sufonylation by heating with arenesulfonyl chlorides in the presence of BiCl3 (5 mol%) and triflic acid (10 mol%) to give high yields of mixed diaryl sulfones; neither BiCl3 nor TfOH alone is effective: Tetrahedron Lett., 40, 9233 (1999).
  6. Promotes the chemoselective deprotection of N-Boc amino acids and peptides: Tetrahedron Lett., 47, 389 (2006).
  7. For a brief feature on Bi(III) catalysts, see: Synlett, 1194 (2001). For a review of their use in acylation and sulfonylation reactions, see: Synlett, 181 (2002).
  8. Antonio, M. R.; McAlister, D. R.; Horwitz, E. P. An europium(III) diglycolamide complex: insights into the coordination chemistry of lanthanides in solvent extraction. Dalton Trans. 2015, 44 (2), 515-521.
  9. Vuorinen, S.; Lahcini, M.; Hatanpää, T.; Sundberg, M.; Leskelä, M.; Repo, T. Bismuth(III) Alkoxide Catalysts for Ring-Opening Polymerization of Lactides and ε-Caprolactone. Macromol. Chem. Phys. 2013, 214 (6), 707-715.