9-Bromofluorene, 98+%, Thermo Scientific Chemicals
9-Bromofluorene, 98+%, Thermo Scientific Chemicals
9-Bromofluorene, 98+%, Thermo Scientific Chemicals
9-Bromofluorene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Bromofluorene, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number ALFA10428.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13472-36-1
IUPAC Nametetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular FormulaH20Na4O17P2
InChI KeyVZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH9.5 to 10.5 (5% solution at 25.0°C)
Insoluble matter=<0.01 %
Sulfate (SO4)=<0.005 %
Nitrogen compounds (as N)=<0.001 %
Titration with HCl99.0 to 103.0 %
View more
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
RUO – Research Use Only
General References:
  1. CP Defeo. Erythema multiforme bullosum caused by 9-bromofluorene. Arch Dermatol.196694,(5) , 545-551.
  2. G Nie.; T Cai.; L Qu.; J Xu.; Q Wei.; S Zhang. A novel polyfluorene derivative electrodeposited by direct anodic oxidation of 9-bromofluorene in boron trifluoride diethyl etherate and trifluoroacetic acid. Journal of Electroanalytical Chemistry. 2008612,(2) , 191-200.