Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
CAS: 13446-18-9 | H12MgN2O12 | 256.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFA10329.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Boiling Point330°C (decomposition)
Chemical Name or MaterialMagnesium nitrate hexahydrate
Melting Point89°C
CAS13446-18-9
Health Hazard 1Danger
View more
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
Solubility
Very soluble in water, alcohol and liquid ammonia.
Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
RUO – Research Use Only
General References:
  1. Sharma, A.; Tyagi, V. V.; Chen, C. R.; Buddhi, D. Review on thermal energy storage with phase change materials and applications. Renewable Sustainable Energy Rev. 2009, 13, 318-345.
  2. Sulaiman, M.; Rahman, A. A.; Mohamed, N. S. Structural, Thermal and Conductivity Studies of Magnesium Nitrate - Alumina Composite Solid Electrolytes Prepared via Sol-Gel Method. Int. J. Electrochem. Sci. 2013, 8, 6647 - 6655.