3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminophthalic acid hydrochloride dihydrate, 96%, Thermo Scientific Chemicals
CAS: 1852533-96-0 | C8H8ClNO4 | 217.605 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFA10319.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Acidity/alkalinity=<0.0005 meq/g
Calcium (Ca)=<1 ppm
Iron (Fe)=<1 ppm
Magnesium (Mg)=<0.5 ppm