N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals
N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals
N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals
N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Hydroxysuccinimide, 98+%, Thermo Scientific Chemicals
CAS: 6066-82-6 | C4H5NO3 | 115.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA10312.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Hydroxysuccinimide
CAS6066-82-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point94°C to 99°C
View more
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Hydroxysuccinimide is used for improved amidations in the carbodiimide method. It is also used to activate a carboxyl group and reacts with amine to form amide. It is involved in the preparation of N-hydroxymaleimide-styrene copolymer. Further, it finds use in analytical chemistry. As an additive, it is used in the carbodiimide method for improved amidations and peptide couplings.
Solubility
Soluble in water, dimethyl formamide, alcohols and ethyl acetate. Insoluble in cold ether.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and strong bases. Store in a cool place.
RUO – Research Use Only
General References:
  1. Reagent for the preparation of active N-hydroxysuccinimide esters for peptide coupling with minimum racemization: J. Am. Chem. Soc., 86, 1839 (1964), or as an additive in the DCC method (see N,N'-Dicyclohexyl carbodiimide, A10973 ) to suppress racemization: J. Am. Chem. Soc., 89, 7151 (1967). For peptide reagents, see Appendix 6.
  2. For a study of the aminolysis of N-hydroxysuccinimide esters, see: J. Org. Chem., 53, 3583 (1988).
  3. Park, C.; Vo, C. L. N.; Kang, T.; Oh, E.; Lee, B. J. New method and characterization of self-assembled gelatin-oleic nanoparticles using a desolvation method via carbodiimide-hydroxysuccinimide (EDCHS) reaction. Eur. J. Pharm. Biopharm. 2015, 89, 365-373.
  4. Peng, Z.; McGee, W. M.; Bu, J.; Barefoot, N. Z.; McLuckey, S. A. Gas phase reactivity of carboxylates with N-hydroxysuccinimide esters. J. Am. Soc. Mass. Spectrom. 2015, 26 (1), 174-180.