Biphenyl, 99%, Thermo Scientific Chemicals
Biphenyl, 99%, Thermo Scientific Chemicals
Biphenyl, 99%, Thermo Scientific Chemicals
Biphenyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Biphenyl, 99%, Thermo Scientific Chemicals
CAS: 92-52-4 | C12H10 | 154.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10265.30250 g
Catalog number ALFA10265.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1907-33-1
IUPAC Namelithium(1+) 2-methylpropan-2-olate
Molecular FormulaC4H9LiO
InChI KeyLZWQNOHZMQIFBX-UHFFFAOYSA-N
SMILES[Li+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (special)≥99.0% as LiOC₄H₉
Trace MetalNa: ≤75ppm
Chloride content≤300ppm
Trace MetalBa: ≤1ppm
View more
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds
Solubility
Soluble in alcohol and ether
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.
RUO – Research Use Only
General References:
  1. J. Trotter.The crystal and molecular structure of biphenyl. Acta Cryst. 1961, 14,1135-114.
  2. Detlef E. Schulz; Gert Petrick; Jan C. Duinker.Complete characterization of polychlorinated biphenyl congeners in commercial Aroclor and Clophen mixtures by multidimensional gas chromatography-electron capture detection. Environ. Sci. Technol. 1989, 23, (7), 852-859.
  3. For use in combination with Li metal for the generation of activated Ca metal, see: Org. Synth. Coll., 9, 9 (1998).