Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
CAS: 6381-59-5 | C4H12KNaO10 | 282.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10163.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS148547-19-7
IUPAC Namemethyl 4-bromo-3-methylbenzoate
Molecular FormulaC9H9BrO2
InChI KeyGTZTYNPAPQKIIR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point38°C to 44°C
Appearance (Form)Low melting solid or liquid
GC>=94.0 %
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Baking powder, medicine (cathartic), component of Fehling′s solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
Solubility
Soluble in water. Insoluble in alcohol
Notes
Loses water of crystallization at 140°. Unstable above 225°. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Jalal, T. K.; Ahmed, I. A.; Mikail, M.; Momand, L.; Draman, S.; Isa, M. L. M.; Rasad, M. S. B. A.; Omar, M. N.; Ibrahim, M.; Wahab, R. A. Evaluation of Antioxidant, Total Phenol and Flavonoid Content and Antimicrobial Activities of Artocarpus altilis (Breadfruit) of Underutilized Tropical Fruit Extracts. Appl. Biochem. Biotechnol. 2015, 175 (7), 3231-3243.
  2. Wu, J.; Hothersall, H.; Mazzetti, C.; O'Connell, Y.; Shields, J. A.; Rahman, A. S.; Cox, R. J.; Crosby, J.; Simpson, T. J.; Thomas, C. M.; Willis, C. L. In vivo mutational analysis of the mupirocin gene cluster reveals labile points in the biosynthetic pathway: the leaky hosepipe mechanism. ChemBioChem 2008, 9 (9), 1500-1508.