1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 98%, Thermo Scientific Chemicals
CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10154.30250 g
Catalog number ALFA10154.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS8004-87-3
IUPAC Namebis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Molecular FormulaC24H28N3
InChI KeyHJPVVBCICYOONZ-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Absorption Ratio P-15/P+151 to 1.25
Appearance (Color)Green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrophotometry>=75 % (on dry substance)
View more
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
RUO – Research Use Only
General References:
  1. S M Sami; R T Dorr; D S Alberts; W A Remers. 2-substituted 1,2-dihydro-3H-dibenz[de,h]isoquinoline-1,3-diones. A new class of antitumor agent. Journal of Medicinal Chemistry. 1993, 36 (6), 765-770.
  2. V T Mazaev; L B Troenkina. [Amino compound stability and transformation in an aqueous environment]. Hygiene and Sanitation. 1981, (10), 91-92.