Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
CAS: 1126-46-1 | C8H7ClO2 | 170.59 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10109.22100 g
Catalog number ALFA10109.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS123-72-8
IUPAC Namebutanal
Molecular FormulaC4H8O
InChI KeyZTQSAGDEMFDKMZ-UHFFFAOYSA-N
SMILESCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3770 to 1.3830 (20°C, 589 nm)