Ethyl salicylate, 99%, Thermo Scientific Chemicals
Ethyl salicylate, 99%, Thermo Scientific Chemicals
Ethyl salicylate, 99%, Thermo Scientific Chemicals
Ethyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl salicylate, 99%, Thermo Scientific Chemicals
CAS: 118-61-6 | C9H10O3 | 166.176 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10101.22100 g
Catalog number ALFA10101.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl salicylate
CAS118-61-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c