Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth(III) chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF089485.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4701-17-1
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6380 to 1.6410 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Appearance (Form)Liquid after melting
GC>=96.0 %
It is used as a catalyst in organic chemistry. It is a good source of bismuth and is used to make other bismuth compounds. Also used as a catalyst and to make other bismuth salts.