Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Barium titanium oxide, 99.7% (metals basis), Thermo Scientific Chemicals
CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ALF088267.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium titanium oxide
Melting Point1620°C
CAS12047-27-7
ColorWhite
Health Hazard 1H302+H332
View more
Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.