Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese, AAS standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088078.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear viscous liquid
Color scale=<10 APHA
GC>=99.0 %
Refractive index1.4700 to 1.4750 (20°C, 589 nm)