Di-n-octyltin oxide, Thermo Scientific Chemicals
Di-n-octyltin oxide, Thermo Scientific Chemicals
Di-n-octyltin oxide, Thermo Scientific Chemicals
Di-n-octyltin oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Di-n-octyltin oxide, Thermo Scientific Chemicals
CAS: 870-08-6 | C16H34OSn | 361.16 g/mol
Have Questions?
Catalog NumberQuantity
ALF071193.1425 g
Catalog number ALF071193.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS150020-64-7
IUPAC Name6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
Molecular FormulaC5H2Cl2N2O2S2
InChI KeyAPRATEMYDZDYJD-UHFFFAOYSA-N
SMILESClC1=C(N2C=CSC2=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)136.5-142.5?C
Assay (Titration ex Chloride)≥96.0 to ≤104.0%