Check Order Status
Magnesium, Ion chromatography standard solution, Specpure™, Mg
+
++2 1000μg/ml
Element Search
Structure Search
Thermo Scientific Chemicals
Magnesium, Ion chromatography standard solution, Specpure™, Mg
+
++2 1000μg/ml
Mg(NO{3}){2} in H{2}O
Have Questions?
Catalog number
ALF047258.AE
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS
16940-66-2
IUPAC Name
sodium boranuide
Molecular Formula
BH4Na
InChI Key
YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES
[BH4-].[Na+]
View more
Specifications
Specification Sheet
Appearance (Color)
White
Appearance (Form)
Powder
Titration Iodometric
>=98.5 %