Check Order Status
Calcium, Ion chromatography standard solution, Specpure™, Ca
+
++2 1000μg/ml
Element Search
Structure Search
Thermo Scientific Chemicals
Calcium, Ion chromatography standard solution, Specpure™, Ca
+
++2 1000μg/ml
Ca(NO{3}){2} in H{2}O |
Have Questions?
Catalog number
ALF047234.AE
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Shelf Life
1 Year 6 Months
Physical Form
Liquid
Quantity
100 mL
Molecular Formula
Ca(NO{3}){2} in H{2}O
MDL Number
MFCD00069184
View more