Hematoxylin hydrate, 80% (dry wt.), water <8%, Thermo Scientific Chemicals
Hematoxylin hydrate, 80% (dry wt.), water &lt;8%, Thermo Scientific Chemicals
Hematoxylin hydrate, 80% (dry wt.), water &lt;8%, Thermo Scientific Chemicals
Hematoxylin hydrate, 80% (dry wt.), water &lt;8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hematoxylin hydrate, 80% (dry wt.), water <8%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF047223.22100 g
Catalog number ALF047223.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS123-30-8
IUPAC Name4-aminophenol
Molecular FormulaC6H7NO
InChI KeyPLIKAWJENQZMHA-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to cream to beige to light brown
Appearance (Form)Powder or crystals
GC>=96.0 %
Hematoxylin and eosin together make up Hematoxylin and eosin stain, one of the most commonly used stains in histology. Metal-haematein complexes are used to stain cell nuclei prior to examination under a microscope. Structures that stain with iron- or aluminum-haematein are often called basophilic, even though the mechanism of the staining is different from that of staining with basic dyes.