Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
CAS: 7782-42-5 | C | 12.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF047132.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 %
UVat 270 nm A: =<1
UVat 275 nm A: =<0.3
UVat 295 nm A: =<0.1
UVat 310 nm A: =<0.05