Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
CAS: 133395-16-1 | C4F12MgN2O8S4 | 584.576 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALF046667.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium bis (trifluoromethylsulfonyl)imide
CAS133395-16-1
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.