Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silicon powder, -325 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF046654.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS15764-16-6
IUPAC Name2,4-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyGISVICWQYMUPJF-UHFFFAOYSA-N
SMILESCC1=CC=C(C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥90.0%
Refractive Index1.5095-1.5895 @ 20?C
FormLiquid
Free acid (titration)≤5%