Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
CAS: 124-41-4 | CH3NaO | 54.024 g/mol
Have Questions?
Catalog NumberQuantity
ALF046585.0F2500 mL
Catalog number ALF046585.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium methoxide
CAS124-41-4
Health Hazard 1H226-H301+H311+H331-H314-H335-H370
Health Hazard 2GHS H Statement
H311-H331-H370-H314-H226
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Sodium methoxide 2% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.