Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF046314.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point∼1890°C
Chemical Name or MaterialBismuth (III) oxide
Melting Point817°C
CAS1304-76-3
ColorYellow
View more
Bismuth(III) oxide is used in the preparation of BiFeO3perovskite nanoparticles. It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generators. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.