Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF046314.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red-brown
Appearance (Form)Solution
Titration with NaOH55.0 to 58.0 %
Heavy metals (as Pb)=<10 ppm
Residue after ignition=<0.01 %
View more
Bismuth(III) oxide is used in the preparation of BiFeO3perovskite nanoparticles. It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generators. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.