Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Plant growth hormone
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF045569.09
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS629-97-0
IUPAC Namedocosane
Molecular FormulaC22H46
InChI KeyHOWGUJZVBDQJKV-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White
Appearance (Form)Crystalline solid or crystals and/or chunks
Infrared spectrumConforms
Melting point43°C to 47°C
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.