Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gibberellic acid, 98%, Thermo Scientific Chemicals
Plant growth hormone
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number ALF045569.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS104-94-9
IUPAC Name4-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyBHAAPTBBJKJZER-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56-61?C
FormCrystals or powder or crystalline powder or flakes or granules
Identification (FTIR)Conforms
Appearance (Color)Cream to dark brown or grey or grey/brown
Assay (GC)≥98.5%
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.