Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number ALF045478.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbon
CAS7440-44-0
Health Hazard 1H500
ColorBlack