Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
Thermo Scientific Chemicals
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
CAS: 7782-42-5 | C | 12.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF045365.KS1 Ea.Cardboard Box
Catalog number ALF045365.KS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Ea.
Packaging:
Cardboard Box
Request bulk or custom format
Chemical Identifiers
CAS630-18-2
IUPAC Name2,2-dimethylpropanenitrile
Molecular FormulaC5H9N
InChI KeyJAMNHZBIQDNHMM-UHFFFAOYSA-N
SMILESCC(C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Refractive Index1.3755-1.3795 @ 20?C
FormLiquid
Assay (GC)≥98.0%
Toray Carbon Paper, PTFE treated, TGP-H-60 is used as a gas diffusion layer in fuel cell applications. It is suitable for use as a catalyst backing layer.