Toray Carbon Paper, TGP-H-60, 19x19cm
Toray Carbon Paper, TGP-H-60, 19x19cm
Toray Carbon Paper, TGP-H-60, 19x19cm
Toray Carbon Paper, TGP-H-60, 19x19cm
Thermo Scientific Chemicals
Toray Carbon Paper, TGP-H-60, 19x19cm
CAS: 7782-42-5 | C | 12.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF045356.KS1 Ea.Cardboard Box
Catalog number ALF045356.KS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Ea.
Packaging:
Cardboard Box
Request bulk or custom format
Specifications
Chemical Name or MaterialToray Carbon Paper
CAS7782-42-5
Recommended StorageAmbient temperatures
EINECS Number231-955-3
TSCAYes
View more
Toray Carbon Paper, TGP-H-60 is used as a gas diffusion layer in fuel cell applications. It is suitable for use as a catalyst backing layer.