Manganese powder, APS <10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
Manganese powder, APS &lt;10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
Manganese powder, APS &lt;10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
Manganese powder, APS &lt;10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese powder, APS <10 micron, 99.6% (metals basis), Thermo Scientific Chemicals
CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Catalog NumberQuantity
ALF045098.30250 g
Catalog number ALF045098.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS489-01-0
IUPAC Name2,6-di-tert-butyl-4-methoxyphenol
Molecular FormulaC15H24O2
InChI KeySLUKQUGVTITNSY-UHFFFAOYSA-N
SMILESCOC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals
Melting point102°C to 106°C