Sulfuric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Sulfuric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Sulfuric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Sulfuric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number ALF045048.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
AppearanceColourless solution
Chemical Name or MaterialSulfuric acid
ColorColorless
CAS7664-93-9
CAS7732-18-5