Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
CAS: 11140-77-5 | H40N10O41W12 | 3042.429 g/mol
Have Questions?
Catalog NumberQuantity
ALF044588.0910 g
Catalog number ALF044588.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium tungsten oxide
CAS11140-77-5
Recommended StorageAmbient temperatures
Physical FormPowder
Quantity10 g
View more
Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.