Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF044556.22100 g
Catalog number ALF044556.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS76-60-8
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H14Br4O5S
InChI KeyFRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILESCC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific extinction E (1%/1cm)>=550 (at lambda max)
UV Lambda max613 to 621 nm (in buffer)
Visual transition intervalfrom pH 3.8 (yellow) to pH 5.4 (blue)
Appearance (Color)Yellow to beige to light brown