Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cerium(IV) isopropoxide, Ce 37-45%, Thermo Scientific Chemicals
CAS: 63007-83-0 | C12H28CeO4 | 376.47 g/mol
Have Questions?
Catalog NumberQuantity
ALF044468.065 g
Catalog number ALF044468.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS5029-67-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
FormLow melting solid
Appearance (Color)Colorless to yellow
Cerium(IV) isopropoxide, Ce 37-45% is used to catalyzes diastereoselective pinacol couplings. It forms diketonates ,which hydrolyze to CeO2 particles. CeO2 particles have the potential for ion conduction.