Polystyrene, atactic, Thermo Scientific Chemicals
Polystyrene, atactic, Thermo Scientific Chemicals
Polystyrene, atactic, Thermo Scientific Chemicals
Polystyrene, atactic, Thermo Scientific Chemicals
Polystyrene, atactic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polystyrene, atactic, Thermo Scientific Chemicals
Used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.
Have Questions?
Catalog NumberQuantity
ALF044416.1850 g
Catalog number ALF044416.18
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS78-50-2
IUPAC Name1-(dioctylphosphoryl)octane
Molecular FormulaC24H51OP
InChI KeyZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILESCCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)50-57?C
Identification (FTIR)Conforms
Appearance (Color)White to cream or pale yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or flakes and/or fused solid/chunks
Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.