Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
For determination of Cu and protein assays
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number ALF044247.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS1074-82-4
IUPAC Namepotassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
Molecular FormulaC8H4KNO2
InChI KeyFYRHIOVKTDQVFC-UHFFFAOYSA-M
SMILES[K+].O=C1[N-]C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=98.5 % (On dried substance)
Water=<0.5 % (K.F.)
Appearance (Color)White to yellow to green
For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.