Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 | Er | 167.26 g/mol
Have Questions?
Catalog NumberQuantity
ALF044169.065 g
Catalog number ALF044169.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤10 ( at time of packaging)
Water Content (Karl Fischer Titration)≤0.1%
Residue on Evaporation≤0.002%
Appearance (Color)Clear, colorless to pale yellow
CommentPermanganate reducing substances: Passes Test
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only