Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
CAS: 814-87-9 | C6Al2O12 | 318.02 g/mol
Have Questions?
Catalog NumberQuantity
ALF044149.0910 g
Catalog number ALF044149.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS6381-79-9
IUPAC Nametetrapotassium trihydrate dicarbonate
Molecular FormulaC2H6K4O9
InChI KeyWWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILESO.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.01 %
Phosphate (PO4)=<0.001 %
Heavy metals (ICP-OES)=<5 ppm
Titration with HCl98.5 to 101.0 %
Sulfur compounds (as SO4)=<0.004 %
View more
Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.